Fluorescein Disodium Salt

Supplier: ALDON CORP SE

FF0240-100G FF0240-25G
470301-102EA 72.45 CAD
470301-102 470301-104
Fluorescein Disodium Salt
Chemicals for Science Education

CAS Number: 518-47-8
Formula Weight: 376.28
Formula: C20H10O5Na2
Boiling Point (°C): Decomposes
Freezing Point (°C): Decomposes
Solubility: Water
Synonyms: Acid Yellow 73, C.I. No. 45350, Uranine
Shelf Life (months): 36
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR