Flint Standard Wall Glass Tubings
Supplier: GSC International
5MMSLT-48-5 6MMSLT-48-5 7MMSLT-48-5
470235-752CS 276.5 CAD
470235-752 470235-754 470235-756
Flint Standard Wall Glass Tubings
Tubing Transfer Tubing
This soft-glass tubing is lead and potash free.
  • 5 or 25 lbs. packages available
  • Fire-polished
  • Lead and potash-free
  • Uniform diameter and wall thickness
  • All sizes square cut
The machine-made tubing is designed from a formula that combines all the properties necessary for easy bending and blowing work.
The fire-polished tubing has uniform diameter and wall thickness and is square cut in all sizes.