Glass Vials with Screw Caps
Supplier: Glass Vials
V1545C-FM V2170C-FM
470151-614CS 537.85 CAD
470151-614 470151-622 470157-602 470151-286
Glass Vials with Screw Caps
Vials
Ideal for a variety of uses.
  • Accompanied by a polypropylene foam lined closed top cap
  • Cap designed with a continuous thread 'CT' finish
  • Creates a positive seal
  • Available in a wide range of sizes
Display specimens or keep your science supplies organized with these airtight glass vials featuring polypropylene caps.