Flint Glass Gas Measuring Tubes
61911-00 61911-08
470005-524EA 53.1 CAD
470005-524 470104-262
Flint Glass Gas Measuring Tubes
Tubes Gas Sampling Tubes
These fine flint glass tubes measure the volume of gas generated and absorbed by substances undergoing chemical action.
The top is closed and the tube is graduated from the top downward.