Fizz Wizard and Jammin’ Jelly Reaction
Supplier: UNIVERSE OF SCIENCE INC SE
2100-B
470145-532CS 605.1 CAD
470145-532
Fizz Wizard and Jammin’ Jelly Reaction
Educational Materials Chemistry Educational Materials Consumer Chemistry Learning Activities
Chemistry experiments you can drink and eat. Make soda pop and make it change colors.
Pull tasty jelly things from a solution. Make Polymer Pudding. Learn about chemical reactions, acidbase reactions, indicators, pH and gels. Note: This kit contains food grade materials.
Kit contains: Magic Color Crystals (cabbage extract), sodium bicarbonate, citric acid, calcium acetate, sodium alginate, pipet, spoon, small scoops, cups, stirrers and instructions.
Caution: Do not attempt these experiments with any other chemical samples.