Fire Extinguisher, Max Power Dry Chemical
Supplier: DIVAL SAFETY & SUPPLIES SE
11214B
470122-246EA 208 CAD
470122-246
Fire Extinguisher, Max Power Dry Chemical
First Aid Supplies Fire Extinguishers
This fire extinguisher works on Class B and C fires.
  • Superior proprietary Purple K agent based on potassium bicarbonate is 2.5 times more effective than regular dry chemical agents
  • Aluminum shell
  • Pressure gauge and strap bracket included
LxW: 19.8x4.5"
Charge weight: 5 lbs.
Unit weight: 9.4 lbs.