Fire Extinguisher, Halotron Type

Supplier: LOGISTICS SUPPLY SE
24567B
470122-244EA 593.2 CAD
470122-244
Fire Extinguisher, Halotron Type
First Aid Supplies Fire Extinguishers
Class A, B, and C fires.

  • Inert cooling and smothering agent leaves no residue
  • Effective in protecting environments with sensitive electronic computer equipment
  • Halotron 1 filling is an ozone-friendly replacement for the well-known Halon 1211 material
  • Steel shell, pressure gauge and wall bracket

HxØ: 15 ¼x4 ¼"
Charge weight: 5 lbs.
Unit weight: 10 lbs.

Certifications: EPA approved
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR