Fine-Point Forceps With Guide Pins
Supplier: Dunrite Instruments
140531 140541
470005-440BX 334.95 CAD
470005-440 470007-560
Fine-Point Forceps With Guide Pins
Forceps
Slip proof fine point forceps.
  • Guide pins keep tip aligned
  • Prevents slipping and twisting when closed
  • Textured tips improve hold on slippery material
Used in our own labs, these forceps feature a built-in guide pin to keep the fine tips aligned, which is important for grasping individual vessels or nerves. Stainless steel. Length: 4¹/₂".
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.