Fehling's Solution B (Alkaline)
Supplier: ALDON CORP SE
CAS Number: Mixture
Formula Weight: Mixture
Formula: Mixture
Hazard Info: Caustic, Corrosive
Density (g/mL): Approximately 1.1
Boiling Point (°C): Approximately 100
Freezing Point (°C): Approximately 0
Synonyms: Fehling's Alkaline Tartrate Solution
Shelf Life (months): 36
Storage: White
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...