Ward's® Exploring Mechanical Weathering Lab Activity
Supplier: Avantor
470014-594KT 109.8 CAD
470014-594 470023-720 470328-390
Ward's® Exploring Mechanical Weathering Lab Activity
Educational Classroom Kits and Activities
The Exploring Mechanical Weathering Lab Activity Shows the weathering of rocks on the Earth’s surface.
  • Students observe how the environment effects rocks
  • Developed by Ward's geologists
  • Teacher's guide included
  • Aligned to NGSS standards (MS/HS: ESS1, ESS2)
It’s easy to recreate the process of mechanical weathering in the lab. Students can observe what happens to different rock samples when each is shaken and abraded in a container of water.
Their findings help them understand the various factors that influence mechanical weathering. The lab includesMore Product Information