Ward's® Exploring Chemical Weathering Lab Activity
Supplier: Avantor
470014-596KT 105.2 CAD
470014-596 470328-392
Ward's® Exploring Chemical Weathering Lab Activity
Educational Classroom Kits and Activities
Extended Study of the Effects of Weathering.
  • Demonstrate how the chemical environment effects rocks
  • Developed by Ward's® geologists
  • Aligned to NGSS standards (MS/HS: ESS2)
How do chemicals in the environment affect the weathering of the Earth’s rocks? Students find out for themselves by exposing four different rock samples to a dilute acid and measuring the effects over several days.
It comes with a teacher’s guide and student copymasters.
The kit is also available for purchase with a link to a NewPath digital lesson, which includes a variety ofMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable