Eudiometer
Supplier: VWR International
470219-674 503-2-10
470219-674CS 2509.7 CAD
470219-674 470235-712
Eudiometer
Educational Materials Chemistry Educational Materials Properties of Matter Learning Activities in Chemistry
Demonstrate the combining volume ratios of gases.
  • Two sizes available
  • Platinum electrodes included
Long, sturdy glass tubes have platinum electrodes sealed at the closed end with a 2 mm arc gap. The tubes are marked with a permanent brown scale in 0.1 or 0.2 mL divisions for 50 and 100 mL respectively. An induction coil is recommended for use with eudiometers.