Ethylenediamine
Supplier: ALDON CORP SE
Boiling Point: (°C): 117–118
CAS Number: 107-15-3
Density (g/mL): 0.898
Formula: H2NCH2CH2NH2
Formula Weight: 78.12
Freezing Point (°C): 11
Hazard Info: Flammable, Irritant, Toxic, Corrosive
Shelf Life (months): 36
Solubility: Water and Alcohol
Storage: Red
Please Note: This product is designed for educational and teaching laboratories and no certificate of analysis is available.
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...