Ethyl Ether

Supplier: ALDON CORP SE

DD0037-500MLR DD0037-500ML DD0037-100MLR
470301-080EA 78.3 CAD
470301-080 470301-076 470301-078
Ethyl Ether
Chemicals for Science Education

CAS Number: 60-29-7
Formula Weight: 74.12
Formula: (C2H5)2O
Hazard Info: Flammable, Explosive, Toxic
Density (g/mL): 0.71
Boiling Point (°C): 35.8
Freezing Point (°C): -123
Solubility: Common Organic Solvents and Slightly in Water
Synonyms: Diethyl Ether, Ether
Shelf Life (months): 12
Storage: Red


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR