Ethyl Alcohol Denatured

Supplier: ALDON CORP SE

EE0076-20L EE0069-3.8L EE0076-CS/2 EE0076-3.8L EE0076-500ML EE0076-3.8LR EE0075-1LR EE0075-500MLR EE0076-1L EE0076-3.8SC EE0076-PK/2 EE0076-3.8SCR EE0076-500MLR EE0075-3.8SCR EE0075-20LR EE0075-3.8LR
470108-538EA 359.35 CAD
470108-538 470301-044 470301-052 470301-054 470301-058 470301-062 470301-068 470301-074 470301-046 470301-050 470301-056 470301-060 470301-064 470301-066 470301-070 470301-072
Ethyl Alcohol Denatured
Chemicals for Science Education

CAS Number: Mixture
Formula: Mixture
Density: 0.794 g/mL
Boiling and Freezing Point: 78°C, -114°C
Solubility: Miscible with Water and Organic Solvents
Synonyms: Denatured Alcohol, Ethyl Alcohol Anhydrous, Ethanol
Shelf Life: 36 Months


  • Aligned with GLOBE protocols


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.


Caution: This product is flammable and toxic.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR