Ethyl Acetate

Supplier: ALDON CORP SE

EE0068-1L EE0068-3.8SC EE0068-500ML EE0068-100MLR EE0068-3.8SCR EE0068-500MLR
470301-032EA 86.5 CAD
470301-032 470301-034 470301-036 470301-038 470301-040 470301-042
Ethyl Acetate
Chemicals for Science Education

CAS Number: 141-78-6
Formula Weight: 88.11
Formula: CH3COOC2H5
Hazard Info: Flammable, Explosive, Irritant
Density (g/mL): 0.902
Boiling Point (°C): 77
Freezing Point (°C): -83.6
Solubility: Common Organic Solvents and Slightly in Water
Synonyms: Ethyl Acetic Ester
Shelf Life (months): 36
Storage: Red


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR