Essential Physics Demo: Large Demonstration Lenses
Supplier: Wards
LS125-50 LS125-100 LS125-150 LNX125-50 LNX125-100 LNX125-150 LNSP125-V10 LN-5SPL LN-SPH12
470017-152EA 30 CAD
470017-152 470017-154 470017-156 470017-158 470017-160 470017-162 470017-164 470017-166 470134-818
Essential Physics Demo: Large Demonstration Lenses
Educational Materials Physics Educational Materials Optics Learning Activities
Makes demonstrations viewable by the entire classroom.
  • Large scale for easy demonstrations
  • Parts are available separately
  • Easily demonstrates real and virtual images
Make all your optics experiments easily visible by the entire class with these large demonstration lenses which are easily mounted on a large lens support stand. Demonstrate a telescope, microscope or neutralizing lens by mixing all the different lenses. The set includes six lenses and the large lens support stand.