Ward's® Live Escherichia coli Conjugation Strains
Supplier: Avantor
470179-516EA 55.2 CAD
470179-516 470179-518
Ward's® Live Escherichia coli Conjugation Strains
Specimens Live Specimens Bacteria - Living Material
Two strains of Escherichia coli (donor and host) with complementary sensitivities to Ampicillin and Streptomycin. Watch them trade traits via conjugation.
Conjugation DONOR Culture (F+): Ampicillin Resistant and Streptomycin Sensitive
Conjugation HOST (RECIPIENT) Culture (F-): Ampicillin sensitive and Streptomycin resistant
Ordering information: These products contain living or perishable materials and will ship via overnight or second day delivery. Please specify a delivery date during checkout
Bacterial and fungal cultures are restricted and can only be ordered by educational professionals.
Caution: Although this organism is non pathogenic, Ward’s recommends that you utilize proper aseptic technique when handling all microorganisms.