Eosin Y

Supplier: ALDON CORP SE

EE0055-100ML EE0060-30ML EE0040-5G
470045-612EA 23.7 CAD
470045-612 470301-020 470301-024
Eosin Y
Chemicals for Science Education

CAS Number: 17372-87-1
Formula Weight: 691.88
Formula: C20H6Br4Na2O5
Density (g/mL): 1.018
Boiling Point (°C): Decomposes
Solubility: Water and Alcohol
Synonyms: Acid Red 87, C.I. No. 45380
Shelf Life (months): 36
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR