Ward's® Live Enterobacter aerogenes Culture
Supplier: Avantor
470176-600EA 55.2 CAD
470176-600 470177-254
Ward's® Live Enterobacter aerogenes Culture
Specimens Live Specimens Bacteria - Living Material
This organism is commonly recovered from soil and water. As normal intestinal flora, it can also usually be found in sewage.
  • Biosafety Level: 1
  • Canada: Containment Level 2 - PHAC Licence required
  • Gram Stain: Negative
  • Morphology: Rod
  • Motility: Positive
  • Respiration: Aerobic
All bacterial cultures undergo an extensive quality control program prior to releasing for sale, so they are guaranteed for purity as well as for species.
This organism is offered as a ready-to-use tube culture, as well as in a freeze-dried culture for longer storage.
Freeze-dried organisms come with everything you need to rehydrate and culture the organism when you're ready.
Ordering information: These products contain living or perishable materials and will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Bacterial and fungal cultures are restricted and can only be ordered by educational and research professionals.
Ward's Science is not affiliated with the American Type Culture Collection and does not provide corresponding ATCC numbers for our organisms.
Caution: Though this organism is not considered to be pathogenic, Ward's recommends that you utilize proper aseptic technique when handling all microorganisms.