Ward's® Endospore Staining Kit
Supplier: Ward's
470030-202KT 101.09 CAD
470030-202
Ward's® Endospore Staining Kit
Educational Materials Biology Educational Materials Microbiology Learning Activities
Examine a non-pathogenic bacterial strain that illustrates endospore formation.
  • Demonstration plate culture of Bacillus cereus
Under environmentally stressful conditions, vegetative bacteria enter a heat-resistant stage that helps them survive; this stage is known as an endospore.
The bacterial culture and stain in this kit provide a striking example of this stage.
The kit includes:
A coupon redeemable for a plate culture of Bacillus cereus.
Blank microscope slides and applicator swabs
Staining solutions
Canada: Containment Level 2 – PHAC License required.
Ordering information: This item contains a coupon for live or perishable materials. Please provide a date that the live or perishable components will arrive.
Delivery information: The kit will ship separately from the live component. Kits do not contain perishable components and generally ship via UPS ground. Perishable components will ship via overnight or second day delivery.