Electric Heating Mantles
Supplier: Glas-Col
100A O396 100A O402 100A O406
470202-744EA 516.5 CAD
470202-744 470202-746 470202-748
Electric Heating Mantles
Heaters Heating Mantles
Designed to Operate Below Red Heat at Maximum Input Voltage.
  • Made entirely of glass fabric
  • Hemispherical construction allows full view of flask contents while heating
Mantles provide evenly distributed heat which may be closely controlled by adjustment of the input voltage. (Heating Mantle Controller sold separately WLS-40883 / 470202-750)
With separable 2-wire line cord, 4' L. For 115VAC or DC. CSA listed.
Certifications: CSA listed.