EDTA Disodium Salt

Supplier: ALDON CORP SE

EE0091-100G EE0091-100GR EE0091-25GR EE0097-1L
470301-012EA 37.2 CAD
470301-012 470301-014 470301-016 470225-692
EDTA Disodium Salt
Chemicals for Science Education

CAS Number: 6381-92-6
Formula Weight: 372.24
Formula: C10H14N2Na2O8·2H2O
Solubility: Slightly in Water
Synonyms: Ethylenediaminetetraacetic Acid, Disodium EDTA Dihydrate
Shelf Life (months): 36
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR