Economy Short Stem Funnels, Hutzler
Supplier: Hutzler Manufacturing
808 804 802 804 816
470150-348CS 788.4 CAD
470150-348 470150-350 470150-352 470188-434 470150-346
Economy Short Stem Funnels, Hutzler
Funnels Filtration Funnels Filter Funnels
Funnels are made of durable plastic. Funnels of top inner diameter 70-108 are polypropylene and autoclavable. Funnels with top inner diameter of 127 and 165 are polyethylene and not autoclavable.
The 165 mm size has a wide stem for quick transfers.