Economy Lab Ovens
Supplier: Quincy Labs
30GC 10GC 40GC
470003-786EA 1736.4 CAD
470003-786 470006-402 470123-092
Economy Lab Ovens
Ovens Standard Ovens
Heavy Steel Double-Wall Construction.
  • Includes dual range laboratory thermometer and three wire cord/plug
  • 115 VAC, 50/60 Hz
  • Constructed with UL and CSA listed components
These gravity convection ovens are for drying, baking, sterilizing, evaporating and testing. Maximum temperature is 220°C. Ovens feature corrosive-resistant aluminized steel interior, perforated heat shield for good temperature uniformity and sheathed heating elements for quick run-up and recovery times. Features light gray, hard, scratch-resistant hammer finish with two adjustable chrome plated wire shelves and supports.