Economy Polypropylene Low Form Beakers
Supplier: Maryland Plastics
L-1250 L-1220 L-1260 L-1250
470153-382CS 778.05 CAD
470153-382 470153-386 470149-924 470206-318
Economy Polypropylene Low Form Beakers
Beakers
Ideal for school and kitting use.
  • Griffin Low Form Beakers
  • Inexpensive and Resistant
These affordable polypropylene beakers are unaffected by most acids, alkalis and solvents. The manufacturer describes them as disposable, but most teachers find them tough enough to reuse several times. Autoclavable to 121°C. Designed with a dripless pouring spout and graduations accurate to ±5%.