H-B® Easy Read® Environmentally Friendly, General Purpose Liquid-In-Glass Thermometers
Supplier: Bel-Art Products
B60301-0200 B60301-0300 B60301-0200 B60302-0000
470004-834EA 41.65 CAD
470004-834 470004-838 470123-626 470202-822
H-B® Easy Read® Environmentally Friendly, General Purpose Liquid-In-Glass Thermometers
Thermometers Analog Thermometers
EnviroKleen™ certified thermometers feature black, non-toxic, biodegradable, Enviro-Safe® liquid against lead-free, yellow back glass for easy readability and fewer reading errors.
  • Non-toxic and biodegradable liquid
  • Lead-free
  • Yellow back glass
  • Easy to read
Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
Ordering information: Manufactured and tested by our USA quality team, these thermometers are traceable to NIST and include a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability. Should you require an individual Thermometer Calibration Report, factory calibrated thermometers or custom calibration services are available.