Dropping Bottle with Ground Glass Pipette, 60 ml

Supplier: GSC INTERNATIONAL SE
404-1-10 7-404-1
470020-794EA 7.7 CAD
470338-636 470020-794
Dropping Bottle with Ground Glass Pipette, 60 ml
Bottles Dropping Bottles
The 60 ml flint glass dropping bottle with ground glass pipette can be used in the lab to dispense indicators, stains, or prepared reagents.

  • Bottle with ground glass joint
  • Provided with ground glass pipette
  • 60 ml capacity
  • Delivers 0.05 ml drop

The pipette is designed to deliver a 0.05 ml drop. The bulb is removable and replaceable.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR