Double Sided Mirrors
Supplier: United Scientific Supplies
MSA8X11-F MSA2X3-F MSA4X6-F
470105-954PK 32.65 CAD
470105-954 470105-958 470144-530
Double Sided Mirrors
Mirrors
Flexible mirrors for optical experiments.
  • Acts as a concave or convex mirror
  • Can be cut with scissors
You'll get lots of use out of these double-sided, flexible plastic mirrors that can be used for investigations, observations and creative activities involving reflection, symmetry and more. The mirrors can be cut with scissors.