Disposable Micropipette Tips
Supplier: Edvotek
VWR®
521016 D592 521000 637-2 521014 2249 1903
470149-166CS 99.4 CAD
470149-166 470233-654 470149-174 470148-816 470159-486 470019-832
Disposable Micropipette Tips
Pipette Tips
High quality micropipette tips to meet any lab's needs.
  • Certified DNase, RNase and pyrogen free
  • Material: PP
  • Autoclavable
  • Designed to fit most standard micropipettes
All quantities are supplied in bulk bags.