Dispensing Bottle with Spout Cap
Supplier: Dynalon
605085-0005 605085-01 605085-08
470100-716PK 38.45 CAD
470100-716 470178-162 470202-532
Dispensing Bottle with Spout Cap
Bottles Dropping Bottles
Lightweight Polyethylene.
  • Secure, Snap-On sealer cap
  • Leak-proof seal
Spout openings, 0.03", can be enlarged with a quick snip of scissors.