Direct Reading Anemometer, AMERICAN SCIENTIFIC LLC SE
Supplier: AMERICAN SCIENTIFIC LLC SE
6577-00
470101-782EA 77.3 CAD
470101-782
Direct Reading Anemometer, AMERICAN SCIENTIFIC LLC SE
Anemometers
As the wind blows, the arms on this unique anemometer move and lift the reading bar, allowing your students to get a direct reading of wind speed at just a glance. The colorful plastic instrument can be hand-held or permanently mounted. It measures the Beaufort scale from 0–11. Size: 30 x 18cm.