Ward's® Diorite
Supplier: Avantor
470025-978PK 48.15 CAD
470025-978 470025-980 470025-982
Ward's® Diorite
Specimens Rock Specimens Igneous Rocks
Coarse, “salt and pepper”: dark pyroxene and light andesine.
Ordering information: WARD’S offers different specimen sizes to meet your various needs. Student Specimens are ideal for students to use for study and identification. Student specimens range in size from 1×1" to 1×2" and come in packages of ten. Hand Specimens are great for lectures, demonstrations, and display. Hand specimens range in size from 2×3" to 3×4", and each comes in a labeled plastic tray. Bulk Pack includes several of the smaller pieces that are suitable for classroom study, while the larger specimens in the one kilogram pack can be cut to sizes that meet your specific needs. The number and size of pieces will vary.