Dimethylglyoxime

Supplier: ALDON CORP SE

DD0035-25G DD0035-100GR DD0036-500ML
470300-984EA 39.75 CAD
470300-984 470300-986 470225-716
Dimethylglyoxime
Chemicals for Science Education

CAS Number: 95-45-4
Formula Weight: 116.12
Formula: (CH3C:NOH)2
Hazard Info: Toxic
Freezing Point (°C): 235-240
Solubility: Alcohol and Ether
Synonyms: 2,3-Butanedione dioxime
Shelf Life (months): 36
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR