Diastase of Malt

Supplier: ALDON CORP SE

DD0025-100G DD0025-25G
470300-976EA 54.1 CAD
470300-976 470300-978
Diastase of Malt
Chemicals for Science Education

CAS Number: 9000-92-4
Solubility: Water
Synonyms: Amylase, Malt Diastase, Maltin
Shelf Life (months): 36
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR