Dial Altimeter/Barometer
Supplier: SUN COMPANY, INC.
203-F
470018-694EA 136.25 CAD
470018-694
Dial Altimeter/Barometer
Barometers
Compact, 2-in-1 field tool and weather instrument.
  • Aligned with GLOBE protocols
This great compass companion measures altitude from 0–15,000 feet and barometric pressure from 17–31" Hg. It is protected by a hardside case. Size: 2 3/4" x 3 1/2" x 1".