D-Glucose Monohydrate

Supplier: ALDON CORP SE

DD0021-500G GG0019-37.5G GG0187-500ML GG0180-500ML DD0019-100G GG0111-1L GG0193-7.5G
470301-146EA 30.25 CAD
470301-146 470301-150 470301-152 470301-154 470301-144 470301-148 470301-156
D-Glucose Monohydrate
Chemicals for Science Education

CAS Number: 50-99-7
Formula: C6H12O6
Density: 1.5 g/mL
Freezing Point: 148 °C
Solubility: Water
Synonyms: Dextrose
Shelf Life: 12 Months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR