D-Glucose Anhydrous

Supplier: ALDON CORP SE

DD0019-2.5KG DD0019-500G DD0019-500GR DD0019-500G
470301-138EA 114.3 CAD
470301-138 470301-140 470301-142 470233-594
D-Glucose Anhydrous
Chemicals for Science Education

CAS Number: 50-99-7
Formula: C6H12O6
Density: 1.5 g/mL
Freezing Point: 148 °C
Solubility: Water
Synonyms: Dextrose
Shelf Life: 12 Months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR