D-Fructose

Supplier: ALDON CORP SE

FF0290-25G FF0290-500G FF0290-100G
470300-972EA 20.6 CAD
470300-972 470300-974 470300-970
D-Fructose
Chemicals for Science Education

CAS Number: 57-48-7
Formula Weight: 180.16
Formula: C6H12O6
Density (g/mL): 1.60
Boiling Point (°C): Decomposes
Freezing Point (°C): 104
Solubility: Water, Alcohol, Ether
Synonyms: d-Levulose
Shelf Life (months): 36
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR