Demonstration Electric Meters
Supplier: VWR International
PH1116A
470110-362EA 382.7 CAD
470110-362
Demonstration Electric Meters
Electrical Meters Digital Multimeters
Large, lightweight meters, ideal for classroom demonstrations.
The clearly-visible scales occupy half the visible area of the instruments. To avoid confusion, the scale plate is printed on both sides and is easily reversed, giving only two ranges per scale. A rotary switch on the back to select the measuring range simultaneously changes the measuring units in a window on the front. A zero-adjust knob on the rear shifts the zero point for galvanometer work.
The accuracy of the readings is ±2.5% f.s.d. and the internal resistance of the meter is 49Ω as a galvanometer and 1000Ω/V on the voltage ranges. A pair of test leads and manual are supplied. Meter case measures 28 x 11.5 x 30cm.
Available in Two Versions: The Volts/Ohms/Galvanometer Version has the following ranges: DCV: 1V, 2.5V, 5V, 10V, 25V, 50V, 100V, 150V, 250V; ACV: 10V, 50V, 250V; Ohms: 1Ω - 100Ω, 10Ω - 1kΩ, 100Ω - 10kΩ, 1kΩ - 100kΩ; Galvanometer: -500 – 0 – +500 divisions, 1 A/division. Requires a D-size battery, sold separately. The Volts/Amps/Galvanometer Version has the following ranges: DCV: 5V, 10V; ACV: 10V, 50V, 250V; DCA: 10mA, 100mA, 1A, 5A; ACA: 1A,5A, Galvanometer: -500 – 0 – +500 divisions, 1μA/division.