Demonstration Diffraction Grating
Supplier: VWR International
PH30677/A
470148-784CS 11060.5 CAD
470148-784
Demonstration Diffraction Grating
Educational Materials Physics Educational Materials Optics Learning Activities
This quality card slide lets you demonstrate and compare the properties of light waves.
  • Viewing Area Length: 9 mm
  • Viewing Area Width: 16 mm
  • Gratings Mounted between Glass Plates
  • Three Diffraction Gratings
  • For Demonstrating Properties of Light Waves
It features three different diffraction gratings: 100, 300 and 600 lines per mm.
The gratings are mounted between glass plates.