Ward's® Deluxe Model Stream Table Kits
Supplier: Plas-Labs
105-RT 106-ST 167-RP 168-RP
470029-374EA 2403.35 CAD
470029-374 470029-390 470029-396 470029-398 76628-722 76628-724
Ward's® Deluxe Model Stream Table Kits
Models Stream Models
Stream table kits For the most detailed and thorough demonstrations of sediment transport.
  • Hands-on learning of coastal environments
  • Investigate water movement and how coastlines can change
Demonstrate coastline formation with a wavemaker, follow the course of a stream and more with this table-size stream table kit. Heavy, seamless plastic construction provides years of leakproof use.
Ordering information: Replacement or additional power supplies and pump available for purchase.
Delivery information: Includes a power pack and pump for variable water flow, sediment trap, internal water reservoir, and wave-maker. 125 VAC with 71/2' cord on power pack, 8' cord on pump. The combinations packs also include the rainfall simulator, terrain feature model, contour map grid, and a 32 page stream table investigation model.