DC Motor Kit, Single Kit

Supplier: VWR International
PH1229 DCMKT-12
470003-836EA 65 CAD
470003-836 470111-204
DC Motor Kit, Single Kit
Power Supplies
This kit is a learn-by-building introduction to a motor’s basic components.

  • Focus on Poles, Armatures, and Switches
  • Class Set of 12 or Individual Kits Available
  • One AA Battery Required

Students gain insight into field poles, armatures, and switches.

The kit requires one AA battery, sold separately. It's available individually or as an economical class set of 12.

Ordering information: This kit assembly instructions, a 9 x 6.5 cm mounting base, two coils of insulated wire (200 cm long), a field pole, a mounting bracket, two armature halves, one piece of insulating tubing (4 cm long), a motor shaft (6.5 cm long), two commutator insulators, two bronze wire brushes, two shaft supports, four paper fasteners, a AA battery holder, six screws, and six nuts.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR