Polypropylene, Single Scale Cylinders
Supplier: Bel-Art
F28458-0000_SE F28459-0000
470005-422EA 48.65 CAD
470005-422 470005-424
Polypropylene, Single Scale Cylinders
Cylinders Measuring Cylinders
High Quality, Unbreakable Cylinder.
  • Easy to read scale
  • Autoclavable up to 135°C
Order Now
SPECIFICATIONS
Form Tall
Environmentally Preferable