Cyclohexene

Supplier: ALDON CORP SE

CC0570-100ML CC0570-250ML CC0570-500ML
470300-956EA 69.85 CAD
470300-956 470300-958 470300-960
Cyclohexene
Chemicals for Science Education

CAS Number: 110-83-8
Formula Weight: 82.15
Formula: C6H10
Hazard Info: Flammable, Toxic
Density (g/mL): 0.8102
Boiling Point (°C): 82-84
Freezing Point (°C): -103
Solubility: Alcohol
Synonyms: Benzenetetrahydride, Hexanaphthylene, Tetrahydrobenzene
Shelf Life (months): 12
Storage: Red


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR