Culture Tube Caps
Supplier: DWK Life Sciences (KIMBLE)
73660-13 73660-16
470148-936CS 261.65 CAD
470148-936 470157-376
Culture Tube Caps
Tubes Culture Tubes
Culture tube caps are made of polypropylene.
  • Disposable
  • Autoclavable
  • Packages of 500
Should keep contents free of contamination.