Crucible Tongs
Supplier: United Scientific Supplies
CTSP09 CH13010-3
470005-896EA 14.05 CAD
470005-896 470148-786
Crucible Tongs
Tongs
These tongs are designed for the safe handling of hot objects.
  • Overall length is 9"
  • Riveted joints and serrated tips
Pick up hot crucibles safely. The overall 9" length keeps you at a safe distance ion moving hot crucibles, all while maintaining a controlling grip.