Rescue Critters® Critical Care Fluffy
Supplier: THALES & CO.
210
470029-834EA 3407.9 CAD
470029-834
Rescue Critters® Critical Care Fluffy
Models Zoology Models
A Full-Sized, Realistic Feline Manikin.
  • Perform many emergency procedures on a full size feline manikin
  • Practice restraint, bandaging and venipuncture
  • Made to order
Like its canine counterpart CCJ, Critical Care Fluffy is a full-sized, realistic feline manikin.
It has all the features present in CCJ – trachea, esophagus, epiglottis, tongue, articulated jaw and working lungs. Designed for CPR, veterinarians can perform different techniques in resuscitation on the cat. Moreover, one can also practice cat restraint, bandaging, and venipuncture (there are several vein practice sites.)
CCF accessories include a carrying case, artificial training blood, IV reservoir, IV holder, 5 disposable lungs, endotracheal tube and syringe.
This feline model, ideal for interactive demonstrations of animal care, represents a cat of average weight, with an apical manual pulse.
Ordering information: This item is made to order. Returns cannot be accepted.