Square Glass Coverslips
Supplier: United Scientific Supplies
SCSS18-1OZ SCSS22-1OZ 94-25252
470019-002PK 28.55 CAD
470019-002 470019-004 470151-072 470151-074 470151-078 470145-754
Square Glass Coverslips
Cover Glasses
Coverslip One is 0.15 mm thick. There are approximately 100–115 coverslips per ounce.
  • Non corrosive glass
  • Pre-cleaned
Coverslip Two is 0.2 mm thick. There are approximately 75–100 coverslips per ounce.