Copper(II) Sulfate Pentahydrate

Supplier: ALDON CORP SE
CC0550-2.5KG CC0535-500G CC0535-500GR CC0535-100GRF CC0535-2.5KG CC0535-500GRF CC0545-2.5KG CC0551-50ML CC0535-2.5KGRF CC0535-CAP CC0535-CAP CC0550-500G CC0552-500ML CC0430-CONC CC0545-2.5KGR CC0550-100G CC0535-100GR CC0430-500ML CC0545-500G CC0545-100G CC0545-500GR CC0535-2.5KGR CC0551-500ML
470300-848EA 17.2 CAD
470300-848 470300-858 470300-868 470300-856 470300-878 470300-888 470300-866 470300-854 470300-876 470108-112 470300-864 470300-886 470300-874 470300-896 470300-852 470300-884 470300-862 470300-850 470300-872 470300-894 470300-860 470300-870 470300-892
Copper(II) Sulfate Pentahydrate
Chemicals for Science Education
CAS Number: 7758-99-8
Formula: CuSO4 5H2O
Density: 2.284 g/mL
Freezing Point: 150°C
Solubility: Water and Methanol
Synonyms: Copper II Sulfate 5-Hydrate, Cupric Sulfate 5-Hydrate
Shelf Life: 36 Months

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Caution: This product is a toxic irritant.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR